missing translation for 'onlineSavingsMsg'
Learn More

Trihexylamine 95.0+%, TCI America™

Catalog No. p-7136287
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
T044225ML 25 mL
T0442500ML 500 mL
2 options

Catalog No. T044225ML

Supplier: TCI America T044225ML

Only null left

Chemical Identifiers

CAS 102-86-3
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00009523
InChI Key DIAIBWNEUYXDNL-UHFFFAOYSA-N
Synonym tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
PubChem CID 66022
IUPAC Name N,N-dihexylhexan-1-amine
SMILES CCCCCCN(CCCCCC)CCCCCC

Specifications

Color Yellow
Boiling Point 265°C
Quantity 25 mL
UN Number 3082
Formula Weight 269.52
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trihexylamine