missing translation for 'onlineSavingsMsg'
Learn More

Trihexylamine 95.0+%, TCI America™

Catalog No. T044225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T044225ML 25 mL
T0442500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T044225ML Supplier TCI America Supplier No. T044225ML

Chemical Identifiers

CAS 102-86-3
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00009523
InChI Key DIAIBWNEUYXDNL-UHFFFAOYSA-N
Synonym tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
PubChem CID 66022
IUPAC Name N,N-dihexylhexan-1-amine
SMILES CCCCCCN(CCCCCC)CCCCCC

Specifications

Color Yellow
Boiling Point 265°C
Quantity 25 mL
UN Number 3082
Formula Weight 269.52
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trihexylamine