missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trihexylamine 98.0+%, TCI America™
Supplier: TCI America I036025ML
Specifications
| Trihexylamine [Reagent for Ion-Pair Chromatography] | |
| Colorless | |
| C18H39N | |
| 25 mL | |
| tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl | |
| CCCCCCN(CCCCCC)CCCCCC | |
| 269.517 | |
| 269.52 | |
| Liquid |
| 102-86-3 | |
| 265°C | |
| MFCD00009523 | |
| 3082 | |
| DIAIBWNEUYXDNL-UHFFFAOYSA-N | |
| N,N-dihexylhexan-1-amine | |
| 66022 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 102-86-3 | |
| 269.517 | |
| DIAIBWNEUYXDNL-UHFFFAOYSA-N | |
| 66022 | |
| CCCCCCN(CCCCCC)CCCCCC |
| C18H39N | |
| MFCD00009523 | |
| tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl | |
| N,N-dihexylhexan-1-amine |
Safety and Handling
TSCA : Yes