missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trihexyl(tetradecyl)phosphonium Chloride 97.0+%, TCI America™
$82.79 - $273.38
Chemical Identifiers
| CAS | 258864-54-9 |
|---|---|
| Molecular Formula | C32H68ClP |
| Molecular Weight (g/mol) | 519.32 |
| MDL Number | MFCD04112746 |
| InChI Key | JCQGIZYNVAZYOH-UHFFFAOYSA-M |
| Synonym | Tetradecyl(trihexyl)phosphonium Chloride |
| PubChem CID | 11375816 |
| IUPAC Name | trihexyl(tetradecyl)phosphanium;chloride |
| SMILES | CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC.[Cl-] |
Chemical Identifiers
| 258864-54-9 | |
| 519.32 | |
| JCQGIZYNVAZYOH-UHFFFAOYSA-M | |
| 11375816 | |
| CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC.[Cl-] |
| C32H68ClP | |
| MFCD04112746 | |
| Tetradecyl(trihexyl)phosphonium Chloride | |
| trihexyl(tetradecyl)phosphanium;chloride |
Specifications
| 258864-54-9 | |
| C32H68ClP | |
| 5 g | |
| Tetradecyl(trihexyl)phosphonium Chloride | |
| CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC.[Cl-] | |
| 519.32 | |
| 519.32 | |
| Liquid |
| Colorless | |
| MFCD04112746 | |
| 1760 | |
| JCQGIZYNVAZYOH-UHFFFAOYSA-M | |
| trihexyl(tetradecyl)phosphanium;chloride | |
| 11375816 | |
| ≥97.0% (T) | |
| Trihexyl(tetradecyl)phosphonium Chloride |
Safety and Handling
TSCA : No