missing translation for 'onlineSavingsMsg'
Learn More

Trihexyl Phosphite 95.0+%, TCI America™

Catalog No. P126025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P126025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P126025G Supplier TCI America Supplier No. P126025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6095-42-7
Molecular Formula C18H39O3P
Molecular Weight (g/mol) 334.48
MDL Number MFCD00015282
InChI Key DSKYTPNZLCVELA-UHFFFAOYSA-N
Synonym Phosphorous Acid Trihexyl Ester
PubChem CID 80190
IUPAC Name trihexyl phosphite
SMILES CCCCCCOP(OCCCCCC)OCCCCCC

Specifications

Color Colorless
Boiling Point 158°C
Quantity 25 g
Formula Weight 334.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trihexyl Phosphite