missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triglycerides Reagent

Catalog No. TR22421
Change view
Click to view available options
Quantity:
2 x 125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TR22421 2 x 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TR22421 Supplier Thermo Scientific™ Supplier No. JTR22421
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Determine quantity of triglycerides in human serum or plasma in vitro with Thermo Scientific™ Triglycerides Reagent.

Measurement of triglycerides is important in the diagnosis and management of hyperlipidaemias. Levels may be increased in nephrotic syndrome, coronary artery disease (CAD), hypothyroidism, diabetes mellitus and liver disease. Decreased values are sometimes observed in cases of protein malnutrition, hyperthyroidism and cachectic states.

Specifications

Description Infinity Triglyceride 2 x 125mL
Detectable Analytes Triglycerides
Sample Type Plasma, Serum
Type Reagent
Linearity Up to 10mmol/L (885mg/dL)
Methods Compatibility Endpoint
Quantity 2 x 125 mL
Stability 18 months at 2-8°C