missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluorothymidine 98.0+%, TCI America™
$393.32 - $1163.17
Chemical Identifiers
| CAS | 70-00-8 |
|---|---|
| Molecular Formula | C10H11F3N2O5 |
| Molecular Weight (g/mol) | 296.20 |
| MDL Number | MFCD00006534 |
| InChI Key | VSQQQLOSPVPRAZ-RRKCRQDMSA-N |
| Synonym | trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd |
| PubChem CID | 6256 |
| ChEBI | CHEBI:75179 |
| IUPAC Name | 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2511100MG
|
TCI America
T2511100MG |
100 mg |
Each for $393.32
|
|
|||||
|
T25111G
|
TCI America
T25111G |
1 g |
Each for $1,163.17
|
|
|||||
Chemical Identifiers
| 70-00-8 | |
| 296.20 | |
| VSQQQLOSPVPRAZ-RRKCRQDMSA-N | |
| 6256 | |
| 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C10H11F3N2O5 | |
| MFCD00006534 | |
| trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd | |
| CHEBI:75179 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F |
Specifications
| 70-00-8 | |
| White | |
| MFCD00006534 | |
| trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F | |
| 296.20 | |
| CHEBI:75179 | |
| ≥98.0% (HPLC,T) | |
| Trifluorothymidine |
| 189°C | |
| C10H11F3N2O5 | |
| 100 mg | |
| VSQQQLOSPVPRAZ-RRKCRQDMSA-N | |
| 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 6256 | |
| 296.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XP2087500
TSCA : No