missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluorothymidine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 189°C |
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 296.20 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trifluorothymidine |
Safety and Handling
| RTECSNumber | XP2087500 |
| TSCA | No |