missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)trimethylsilane 97.0+%, TCI America™

Catalog No. p-7136843
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15705G 5 g
T157025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15705G Supplier TCI America Supplier No. T15705G
Only null left

Chemical Identifiers

CAS 81290-20-2
Molecular Formula C5H11F3Si
Molecular Weight (g/mol) 156.22
MDL Number MFCD00145454
InChI Key MTYSDPUZDZURPP-UHFFFAOYSA-N
Synonym trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, Ruppert-Prakash reagent, CF3TMS
PubChem CID 552549
IUPAC Name tert-butyl(trifluoromethyl)silane
SMILES CC(C)(C)[SiH2]C(F)(F)F

Specifications

Color Yellow
Boiling Point 55°C
Quantity 5 g
UN Number 1993
Formula Weight 142.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (Trifluoromethyl)trimethylsilane [Trifluoromethylating Reagent]