missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoromethyl)trimethylsilane, 0.5M soln. in THF

CAS: 81290-20-2 | C5H11F3Si | 156.22 g/mol

Supplier:  Thermo Scientific Chemicals H31520AC

 View more versions of this product

Catalog No. AAH31520AC


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reagent used in nucleophilic addition of the trifluoromethyl group to aldehydes and ketones.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/ moisture.
Specifications

Specifications

40°C
81290-20-2
C5H11F3Si
MFCD00145454
17°C (63°F)
Hydrolyzes in water.
CC(C)(C)[SiH2]C(F)(F)F
156.22
142.2
Moisture sensitive
(Trifluoromethyl)trimethylsilane
25 mL
CF3Si(CH3)3
UN1993
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
MTYSDPUZDZURPP-UHFFFAOYSA-N
trimethyl(trifluoromethyl)silane
552549
0.5M soln. in THF
0.895

Chemical Identifiers

81290-20-2
156.22
MTYSDPUZDZURPP-UHFFFAOYSA-N
552549
CC(C)(C)[SiH2]C(F)(F)F
C5H11F3Si
MFCD00145454
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent
trimethyl(trifluoromethyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only