missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7136560
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T10275G 5 g
T102725G 25 g
2 options

Catalog No. T10275G

Supplier: TCI America T10275G

Only null left

Chemical Identifiers

CAS 421-83-0
Molecular Formula CClF3O2S
Molecular Weight (g/mol) 168.51
MDL Number MFCD00007451
InChI Key GRGCWBWNLSTIEN-UHFFFAOYSA-N
Synonym trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
PubChem CID 79000
IUPAC Name trifluoromethanesulfonyl chloride
SMILES FC(F)(F)S(Cl)(=O)=O

Specifications

Color Yellow
Boiling Point 31°C
Quantity 5 g
UN Number 3265
Formula Weight 168.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator