missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetyl Triflate 97.0+%, TCI America™
Supplier: TCI America T15315G
Specifications
| Trifluoroacetyl Triflate [Powerful Trifluoroacetylating Reagent] | |
| Yellow | |
| MFCD00011638 | |
| 3265 | |
| FWJGTOABGBFQNT-UHFFFAOYSA-N | |
| trifluoromethanesulfonyl 2,2,2-trifluoroacetate | |
| 2735150 | |
| ≥97.0% (T) |
| 68602-57-3 | |
| C3F6O4S | |
| 5 g | |
| Trifluoroacetyl Trifluoromethanesulfonate | |
| FC(F)(F)C(=O)OS(=O)(=O)C(F)(F)F | |
| 246.08 | |
| 246.08 | |
| Liquid |
Chemical Identifiers
| 68602-57-3 | |
| 246.08 | |
| FWJGTOABGBFQNT-UHFFFAOYSA-N | |
| 2735150 | |
| FC(F)(F)C(=O)OS(=O)(=O)C(F)(F)F |
| C3F6O4S | |
| MFCD00011638 | |
| Trifluoroacetyl Trifluoromethanesulfonate | |
| trifluoromethanesulfonyl 2,2,2-trifluoroacetate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator