missing translation for 'onlineSavingsMsg'
Learn More

(Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers) 98.0+%, TCI America™

Catalog No. T31745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31745G 5 g
T31741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31745G Supplier TCI America Supplier No. T31745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 183266-61-7
Molecular Formula C8H4F3N3O
Molecular Weight (g/mol) 215.135
MDL Number MFCD00593044
InChI Key GVQIQOIKWUOEJP-UHFFFAOYSA-N
PubChem CID 3358778
IUPAC Name 1-(benzotriazol-1-yl)-2,2,2-trifluoroethanone
SMILES C1=CC=C2C(=C1)N=NN2C(=O)C(F)(F)F

Specifications

Melting Point 69°C
Color White
Quantity 5 g
Formula Weight 215.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (Trifluoroacetyl)benzotriazole (mixture of 1H- and 2H- isomers)