Learn More
alpha,alpha,alpha-Trifluoroacetophenone, 99%
CAS: 434-45-7 | C8H5F3O | 174.12 g/mol
$88.48 - $88.48
Chemical Identifiers
| CAS | 434-45-7 |
|---|---|
| Molecular Formula | C8H5F3O |
| Molecular Weight (g/mol) | 174.12 |
| MDL Number | MFCD00000420 |
| InChI Key | KZJRKRQSDZGHEC-UHFFFAOYSA-N |
| Synonym | trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 |
| PubChem CID | 9905 |
| IUPAC Name | 2,2,2-trifluoro-1-phenylethanone |
| SMILES | C1=CC=C(C=C1)C(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148350050
|
Thermo Scientific Chemicals
148350050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 434-45-7 | |
| 174.12 | |
| KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
| 9905 | |
| C1=CC=C(C=C1)C(=O)C(F)(F)F |
| C8H5F3O | |
| MFCD00000420 | |
| trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
| 2,2,2-trifluoro-1-phenylethanone |
Specifications
| 434-45-7 | |
| 100.0 | |
| Colorless to Yellow | |
| 165.0°C to 166.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4580 to 1.4600 | |
| MFCD00000420 | |
| 01,1226; 03,308 | |
| trifluoroacetophenone, 2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-phenylethan-1-one, phenyl trifluoromethyl ketone, trifluoromethyl phenyl ketone, alpha,alpha,alpha-trifluoroacetophenone, ethanone, 2,2,2-trifluoro-1-phenyl, 2,2,2-trifluoro-1-phenyl-ethanone, trifluoroacetylbenzene, unii-6t7l1upy09 | |
| KZJRKRQSDZGHEC-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-phenylethanone | |
| 9905 | |
| 99% | |
| α, α, α-Trifluoroacetophenone |
| 98.5 | |
| -40.0°C | |
| 1.2400g/mL | |
| 41°C | |
| 98.5% min. (GC) | |
| C8H5F3O | |
| C6H5COCF3 | |
| 5 g | |
| 1.24 | |
| Solubility in water: insoluble | |
| C1=CC=C(C=C1)C(=O)C(F)(F)F | |
| 174.12 | |
| 174.12 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Warning
EINECSNumber : 207-103-1
RUO – Research Use Only