missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Anhydride 98.0+%, TCI America™

Catalog No. T043320ML
Change view
Click to view available options
Quantity:
20 mL
100 mL
400 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043320ML 20 mL
T0433100ML 100 mL
T0433400ML 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T043320ML Supplier TCI America Supplier No. T043320ML

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
PubChem CID 9845
IUPAC Name 2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 40°C
Quantity 20 mL
UN Number 3265
Formula Weight 210.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trifluoroacetic Anhydride
EINECSNumber (2)-1187
RTECSNumber AJ9800000
TSCA Yes