missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid 99.0+%, TCI America™
$39.14 - $238.60
Chemical Identifiers
| CAS | 76-05-1 |
|---|---|
| Molecular Formula | C2HF3O2 |
| Molecular Weight (g/mol) | 114.02 |
| MDL Number | MFCD00004169 |
| InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
| Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
| PubChem CID | 6422 |
| ChEBI | CHEBI:45892 |
| IUPAC Name | trifluoroacetic acid |
| SMILES | OC(=O)C(F)(F)F |
Chemical Identifiers
| 76-05-1 | |
| 114.02 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| 6422 | |
| trifluoroacetic acid |
| C2HF3O2 | |
| MFCD00004169 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| CHEBI:45892 | |
| OC(=O)C(F)(F)F |
Specifications
| 76-05-1 | |
| Colorless | |
| MFCD00004169 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| OC(=O)C(F)(F)F | |
| 114.02 | |
| CHEBI:45892 | |
| ≥99.0% (T) | |
| Liquid |
| 73°C | |
| C2HF3O2 | |
| 2699 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid | |
| 6422 | |
| 114.02 | |
| 25 g | |
| Trifluoroacetic Acid |
Safety and Handling
EINECSNumber : (2)-1185
RTECSNumber : AJ9625000
TSCA : Yes