missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid 99.0+%, TCI America™

Catalog No. T0431100G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0431100G 100 g
T043125G 25 g
T0431500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0431100G Supplier TCI America Supplier No. T0431100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Boiling Point 73°C
Color Colorless
Quantity 100 g
Physical Form Liquid
UN Number 2699
Formula Weight 114.02
Percent Purity ≥99.0% (T)
Chemical Name or Material Trifluoroacetic Acid
EINECSNumber (2)-1185
RTECSNumber AJ9625000
TSCA Yes