missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, sodium salt, 97%

Catalog No. p-41283
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157930250 25 g Glass bottle
AC157931000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC157930250 Supplier Thermo Scientific Chemicals Supplier No. 157930250
Only null left

CAS: 2923-18-4 | C2F3NaO2 | 136.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2F3NaO2
Molecular Weight (g/mol) 136.01
MDL Number MFCD00013217
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
PubChem CID 517019
IUPAC Name sodium;2,2,2-trifluoroacetate
SMILES [Na+].[O-]C(=O)C(F)(F)F

Specifications

Melting Point 207.0°C to 209.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula CF3CO2Na
Quantity 25 g
Beilstein 02, II, 186
Solubility Information Solubility in water: 625g/L water (25°C). Other solubilities: 131g/L in cf3cooh (30°C)
Formula Weight 136.01
Percent Purity 0.97
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetic acid, sodium salt, 0.97
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 220-879-6
RTECSNumber AK0250000
TSCA TSCA

RUO – Research Use Only