missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid 99.0+%, TCI America™

Catalog No. Q005325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q005325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q005325ML Supplier TCI America Supplier No. Q005325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Color Colorless
Quantity 25 mL
Physical Form Liquid
UN Number 2699
Formula Weight 114.02
Percent Purity ≥99.0% (T)
Chemical Name or Material Trifluoroacetic Acid [Sequenation Reagent]
EINECSNumber (2)-1185
RTECSNumber AJ9625000
TSCA Yes