missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, Reagents

Catalog No. 01336908
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336908 2.5 L
01336907 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336908 Supplier Labchem Inc Supplier No. C23426602.5Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trifluoroacetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym Perfluoroacetic Acid, Trifluoroethanoic Acid
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

pH <1
Melting Point -15.4°C
Boiling Point 72.4°C
Color Colorless to Slight Yellow
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.535
Grade Reagent
Chemical Name or Material Trifluoroacetic Acid