missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Reagent Grade, 99%, Solstice

Catalog No. 60048201
Change view
Click to view available options
Quantity:
10 Ampules
5 x 10 Ampules
25 mL
100 mL
500 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048201 10 Ampules
6001760 25 mL
6001758 100 mL
6001761 500 mL
6001759 1 L
60048203 5 x 10 Ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60048201 Supplier Solstice Supplier No. T650810AMP

Reagent Grade, 99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Density 1.489g/mL at 20°C
Quantity 10 Ampules
Flash Point >100°C (>212°F)
Linear Formula CF3COOH
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
UN Number UN2699
Formula Weight 114.02
Percent Purity 99%
Chemical Name or Material Trifluoroacetic acid
Hazard Statement P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8 / PGI
RTECSNumber 200-929-3