missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Linear Formula | CF3COOH |
| UN Number | UN2699 |
| Beilstein | 742035 |
| Formula Weight | 114.02g/mol |
| Percent Purity | 99% |
| Grade | ReagentPlus |
| Density | 1.489g/mL at 20°C |
| Quantity | 100 mL |
| Flash Point | >100°C (>212°F) |
| Vapor Density | 3.9 (vs air) |
| Show More |
Safety and Handling
| Hazard Statement | P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| RTECSNumber | AJ9625000 |