missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, OR, Macron Fine Chemicals™

Catalog No. MK1942500
Change view
Click to view available options
500g; NMA glass btl/single shipper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK1942500 500g; NMA glass btl/single shipper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK1942500 Supplier Avantor Supplier No. AVA194259

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 76-05-1
Molecular Formula C2HF3O2
Quantity 500g
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoracetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES C(=O)(C(F)(F)F)O
IUPAC Name 2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 114.023
PubChem CID 6422
ChEBI CHEBI:45892
Formula Weight 114.03
Percent Purity 99% min.
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less