missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, MilliporeSigma™

Catalog No. p-3176329
Change view
Click to view available options
Quantity:
70 mL
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MTX12751 70 mL Glass Bottle
MTX12753 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MTX12751 Supplier Technidata Supplier No. TX12751
Only null left

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15.34°C
Boiling Point 72.46°C
Linear Formula CF3COOH
Formula Weight 114.2
Grade NF/USP
Quantity 70 mL
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Trifluoroacetic Acid
Recommended Storage Room Temperature