missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Eluent additive for LC-MS, Honeywell Fluka™

Catalog No. p-7121631
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
1 L
Packaging:
Ampoule
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046823 10 mL Ampoule
60046824 5 x 10 mL Ampoule
6001753 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046823 Supplier Honeywell Chemicals Supplier No. 4096710ML
Only null left

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm, max. 0.1 at 270 nm, max. 0.9 at 260 nm
Melting Point -15.4°C (literature)
Boiling Point 72°C
CAS Max % 100.0000%
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
IUPAC Name trifluoroacetic acid
Formula Weight 114.02
Percent Purity >99%
Grade LC-MS
Density 1.489 g/mL at 20 °C(lit.)
Quantity 10 mL
Flash Point 100°C
Packaging Ampoule
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid, Eluent additive for LC-MS
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
ShelfLife 360 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: I; Proper Shipping Name: Trifluoroacetic acid
EINECSNumber 200-929-3
RTECSNumber 200-929-3
Recommended Storage Room Temp