missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Eluent additive for LC-MS, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Absorbance | max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm, max. 0.1 at 270 nm, max. 0.9 at 260 nm |
| Melting Point | -15.4°C (literature) |
| Boiling Point | 72°C |
| CAS Max % | 100.0000% |
| Linear Formula | CF3COOH |
| UN Number | UN2699 |
| Beilstein | 742035 |
| IUPAC Name | trifluoroacetic acid |
| Formula Weight | 114.02 |
| Percent Purity | >99% |
| Show More |
Safety and Handling
| Hazard Statement | P273-P280-P305 + P351 + P338-P310 |
| ShelfLife | 360 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: I; Proper Shipping Name: Trifluoroacetic acid |
| EINECSNumber | 200-929-3 |
| RTECSNumber | 200-929-3 |
| Recommended Storage | Room Temp |