missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Eluent additive for LC-MS, Honeywell Fluka™

Catalog No. 6001753
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
1 L
Packaging:
Ampoule
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001753 1 L Glass bottle
60046823 10 mL Ampoule
60046824 5 x 10 mL Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001753 Supplier Honeywell Chemicals Supplier No. 409671L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
IUPAC Name trifluoroacetic acid
Formula Weight 114.02g/mol
Percent Purity ≥99% (GC)
Density 1.489g/mL at 20°C
Quantity 1 L
Packaging Glass bottle
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
RTECSNumber AJ9625000