Learn More
Thermo Scientific Chemicals Trifluoroacetic acid, biochemical grade, 99.5+%
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
$121.64 - $963.86
Chemical Identifiers
| CAS | 76-05-1 |
|---|---|
| Molecular Formula | C2HF3O2 |
| Molecular Weight (g/mol) | 114.02 |
| MDL Number | MFCD00004169 |
| InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
| Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
| PubChem CID | 6422 |
| ChEBI | CHEBI:45892 |
| IUPAC Name | trifluoroacetic acid |
| SMILES | OC(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1436518
|
Thermo Scientific Chemicals
A1436518 |
50 g |
Each for $121.64
|
|
|||||
|
AAA1436530
|
Thermo Scientific Chemicals
A1436530 |
250 g |
Each for $312.47
|
|
|||||
|
AAA143650B
|
Thermo Scientific Chemicals
A143650B |
1000 g |
Each for $963.86
|
|
|||||
Description
Used to cleave peptide blocking groupsThe combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed to cleave peptide blocking groupsThe combination of properties like solubility in most of the solvents, volatility, catalytic property, and strong acidity with non-oxidizing nature makes it a widely used reagent in organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and performance products. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. TFA finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.
Solubility
Miscible with ether, acetone, ethanol, benzene, hexane and carbon tetrachloride.
Notes
Trifluoroacetic acid (TFA) is a stable, hygroscopic, volatile and low-boiling organic acid. Storage in a cool place is recommended.
Chemical Identifiers
| 76-05-1 | |
| 114.02 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| 6422 | |
| trifluoroacetic acid |
| C2HF3O2 | |
| MFCD00004169 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| CHEBI:45892 | |
| OC(=O)C(F)(F)F |
Specifications
| -15°C | |
| 72°C to 73°C | |
| MFCD00004169 | |
| 742035 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid | |
| 6422 | |
| 114.02 | |
| Biochemical | |
| 50 g | |
| 1.284 | |
| Trifluoroacetic acid |
| 76-05-1 | |
| C2HF3O2 | |
| UN2699 | |
| 14,9681 | |
| Miscible with ether,acetone,ethanol,benzene,hexane and carbon tetrachloride. | |
| OC(=O)C(F)(F)F | |
| 114.02 | |
| CHEBI:45892 | |
| ≥99.5% | |
| 1.48 | |
| Pungent | |
| Hygroscopic |
Safety and Handling
GHS H Statement
H314-H332
Causes severe skin burns and eye damage.
Harmful if inhaled.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: TRIFLUOROACETIC ACID
EINECSNumber : 200-929-3
RTECSNumber : AJ9625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only