missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 27 kg |
| Packaging | Drum |
| Percent Purity | 99.9% |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.