missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor TR11925GM

 View more versions of this product

Catalog No. 18614560


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

76-05-1
99.9%
25 g
1
Reagent
Amber Glass Bottle

Chemical Identifiers

76-05-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.