missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical

Catalog No. 18614558
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
27 kg
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614558 100 g Amber Glass Bottle
18614562 500 g Amber Glass Bottle
18614559 1 kg Amber Glass Bottle
18614563 2.5 kg Glass Bottle
18614561 27 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614558 Supplier Spectrum Chemical Mfg Cor Supplier No. TR119100GM

Chemical Identifiers

CAS 76-05-1

Specifications

CAS Max % 1
Percent Purity 99.9%
Grade Reagent
Quantity 100 g
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.