missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 471670025
Specifications
| Trifluoroacetic acid | |
| 76-05-1 | |
| Colorless to light yellow clear liquid | |
| CF3CO2H | |
| 2699 | |
| Miscible in water, miscible with ether, acetone, ethanol, benzene, CCl4, hexane, methanol, xylene | |
| OC(=O)C(F)(F)F | |
| 97.5% to 102.5% | |
| 114.02 | |
| Pure | |
| 2.5 kg | |
| Glass Bottle | |
| 1.3 | |
| 107 mbar (25°C) |
| -15°C | |
| 72°C (760 mmHg) | |
| C2HF3O2 | |
| MFCD00004169 | |
| TFA | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid | |
| 114.02 | |
| 98% | |
| 1.535 | |
| Conforms to structure | |
| Liquid | |
| Hygroscopic |
Chemical Identifiers
| 76-05-1 | |
| 114.02 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid |
| C2HF3O2 | |
| MFCD00004169 | |
| TFA | |
| OC(=O)C(F)(F)F |
Safety and Handling
GHS05: Corrosion
GHS07: Exclamation mark
ShelfLife : 2 years
EINECSNumber : 200-929-3
RTECSNumber : AJ9625000
Recommended Storage : Normal conditions