missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, 98%, Thermo Scientific™

Catalog No. AC471670250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC471670250 25 g
AC471671000 100 g
AC471675000 500 g
AC471670010 1 kg
AC471670025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC471670250 Supplier Thermo Scientific Chemicals Supplier No. 471670250

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym TFA
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C (760 mmHg)
Appearance Colorless to light yellow clear liquid
Linear Formula CF3CO2H
UN Number 2699
Solubility Information Miscible in water, miscible with ether, acetone, ethanol, benzene, CCl4, hexane, methanol, xylene
Concentration 97.5% to 102.5%
Formula Weight 114.02
Percent Purity 98%
Grade Pure
Density 1.535
Quantity 25 g
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3
Sensitivity Hygroscopic
Vapor Pressure 107 mbar (25°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
Hazard Category GHS07: Exclamation mark
Hazard Statement GHS05: Corrosion
ShelfLife 2 years
EINECSNumber 200-929-3
RTECSNumber AJ9625000
Recommended Storage Normal conditions