missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde methyl hemiacetal, tech. 90%

Catalog No. AAA1836906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1836906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1836906 Supplier Thermo Scientific Chemicals Supplier No. A1836906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 431-46-9 | C3H5F3O2 | 130.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 431-46-9
Molecular Formula C3H5F3O2
Molecular Weight (g/mol) 130.07
MDL Number MFCD00013572
InChI Key GWTBCUWZAVMAQV-UHFFFAOYNA-N
Synonym trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal
PubChem CID 9892
SMILES COC(O)C(F)(F)F

Specifications

Density 1.36
Boiling Point 96°C
Flash Point 42°C (107°F)
Refractive Index 1.333
Quantity 5 g
UN Number UN3271
Beilstein 1737758
IUPAC Name 2,2,2-trifluoro-1-methoxyethanol
Formula Weight 130.07
Percent Purity 90%
Grade Technical
Chemical Name or Material Trifluoroacetaldehyde methyl hemiacetal
Show More Show Less
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 207-072-4
RTECSNumber KL6600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only