missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetaldehyde Methyl Hemiacetal (contains ca. 10% Methanol) 88.0+%, TCI America™
Supplier: TCI America T324725G
Specifications
| Trifluoroacetaldehyde Methyl Hemiacetal (contains ca. 10% Methanol) | |
| Colorless | |
| C3H5F3O2 | |
| 25 g | |
| trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal | |
| COC(O)C(F)(F)F | |
| 130.07 | |
| 130.07 | |
| Liquid |
| 431-46-9 | |
| 98°C | |
| MFCD00013572 | |
| 1993 | |
| GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
| 2,2,2-trifluoro-1-methoxyethan-1-ol | |
| 9892 | |
| ≥88.0% (GC) |
Chemical Identifiers
| 431-46-9 | |
| 130.07 | |
| GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
| 9892 | |
| COC(O)C(F)(F)F |
| C3H5F3O2 | |
| MFCD00013572 | |
| trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal | |
| 2,2,2-trifluoro-1-methoxyethan-1-ol |
Safety and Handling
RTECSNumber : KL6600000
TSCA : Yes