missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde Methyl Hemiacetal (contains ca. 10% Methanol) 88.0+%, TCI America™

Catalog No. T32475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32475G 5 g
T324725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32475G Supplier TCI America Supplier No. T32475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 431-46-9
Molecular Formula C3H5F3O2
Molecular Weight (g/mol) 130.07
MDL Number MFCD00013572
InChI Key GWTBCUWZAVMAQV-UHFFFAOYNA-N
Synonym trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal
PubChem CID 9892
IUPAC Name 2,2,2-trifluoro-1-methoxyethan-1-ol
SMILES COC(O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
UN Number 1993
Formula Weight 130.07
Percent Purity ≥88.0% (GC)
Physical Form Liquid
Chemical Name or Material Trifluoroacetaldehyde Methyl Hemiacetal (contains ca. 10% Methanol)
RTECSNumber KL6600000
TSCA Yes