missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde Ethyl Hemiacetal (contains ca. 10% Ethanol) 80.0+%, TCI America™

Catalog No. t079110g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T079110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T079110G Supplier TCI America Supplier No. T079110G

Chemical Identifiers

CAS 433-27-2
Molecular Formula C4H7F3O2
Molecular Weight (g/mol) 144.093
MDL Number MFCD00000441
InChI Key KLXJPQNHFFMLIG-UHFFFAOYSA-N
Synonym trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol
PubChem CID 9897
IUPAC Name 1-ethoxy-2,2,2-trifluoroethanol
SMILES CCOC(C(F)(F)F)O

Specifications

Color Colorless
Quantity 10 g
UN Number 1993
Formula Weight 144.09
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Trifluoroacetaldehyde Ethyl Hemiacetal (contains ca. 10% Ethanol)
RTECSNumber KK9000000
TSCA Yes