missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde ethyl hemiacetal, 90%

Catalog No. p-204641
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC139710050 5 mL Glass bottle
1 options

Catalog No. AC139710050

Supplier: Thermo Scientific Chemicals 139710050

Only null left

CAS: 433-27-2 | C4H7F3O2 | 144.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433-27-2
Molecular Formula C4H7F3O2
Molecular Weight (g/mol) 144.09
MDL Number MFCD00000441
InChI Key KLXJPQNHFFMLIG-UHFFFAOYSA-N
Synonym trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol
PubChem CID 9897
IUPAC Name 1-ethoxy-2,2,2-trifluoroethanol
SMILES CCOC(C(F)(F)F)O

Specifications

Color Colorless
Density 1.2400g/mL
Boiling Point 104°C to 105°C (745.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.3410 to 1.344
Linear Formula CF3CH(OC2H5)OH
Quantity 5 mL
Beilstein 01, IV, 3133
Specific Gravity 1.24
Formula Weight 144.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Trifluoroacetaldehyde ethyl hemiacetal, 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
EINECSNumber 207-087-6
RTECSNumber KK9000000
TSCA TSCA

RUO – Research Use Only