missing translation for 'onlineSavingsMsg'
Learn More

Triflumuron, 98+%, Thermo Scientific Chemicals

Catalog No. p-7042606
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6311222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAJ6311222 Supplier Thermo Scientific Chemicals Supplier No. J6311222
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA

Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64628-44-0
Molecular Formula C15H10ClF3N2O3
Molecular Weight (g/mol) 358.701
MDL Number MFCD00072496
InChI Key XAIPTRIXGHTTNT-UHFFFAOYSA-N
Synonym triflumuron, alsystin, trifluron, mascot, alsystine, triflumuron iso, caswell no. 217c, bay-sir 8514, unii-3ft64dyg8k, bay-vi 7533
PubChem CID 47445
ChEBI CHEBI:39388
IUPAC Name 2-chloro-N-[[4-(trifluoromethoxy)phenyl]carbamoyl]benzamide
SMILES C1=CC=C(C(=C1)C(=O)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl

Specifications

Melting Point 198°C
Quantity 100 g
Beilstein 2776684
Merck Index 14,9678
Solubility Information Soluble in DMF
Formula Weight 358.7
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Triflumuron
EINECSNumber 264-980-3
RTECSNumber CV2474000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only