Learn More
Triethylsulfonium iodide, 97%
CAS: 1829-92-1 | C6H15IS | 246.15 g/mol
Supplier: Thermo Scientific Chemicals L0066714
Description
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
| Triethylsulfonium iodide | |
| ∽148°C (decomposition) | |
| MFCD00051867 | |
| 3911790 | |
| triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide | |
| LKDQWVKWYGOVJW-UHFFFAOYSA-M | |
| triethylsulfanium;iodide | |
| 74589 | |
| 97% |
| 1829-92-1 | |
| C6H15IS | |
| 25 g | |
| Light sensitive | |
| It is soluble in water. | |
| CC[S+](CC)CC.[I-] | |
| 246.15 | |
| 246.15 |
Chemical Identifiers
| 1829-92-1 | |
| 246.15 | |
| LKDQWVKWYGOVJW-UHFFFAOYSA-M | |
| 74589 | |
| CC[S+](CC)CC.[I-] |
| C6H15IS | |
| MFCD00051867 | |
| triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide | |
| triethylsulfanium;iodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-383-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only