missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium iodide, 97%

Catalog No. AAL0066722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0066722 100 g
AAL0066706 5 g
AAL0066714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0066722 Supplier Thermo Scientific Chemicals Supplier No. L0066722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1829-92-1 | C6H15IS | 246.15 g/mol

Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1829-92-1
Molecular Formula C6H15IS
Molecular Weight (g/mol) 246.15
MDL Number MFCD00051867
InChI Key LKDQWVKWYGOVJW-UHFFFAOYSA-M
Synonym triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide
PubChem CID 74589
IUPAC Name triethylsulfanium;iodide
SMILES CC[S+](CC)CC.[I-]

Specifications

Melting Point ∽148°C (decomposition)
Quantity 100 g
Beilstein 3911790
Sensitivity Light sensitive
Solubility Information It is soluble in water.
Formula Weight 246.15
Percent Purity 97%
Chemical Name or Material Triethylsulfonium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-383-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only