missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™
Supplier: TCI America T168925G
Specifications
| Triethylsilyl Trifluoromethanesulfonate | |
| Yellow | |
| C7H15F3O3SSi | |
| 25 g | |
| triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf | |
| CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F | |
| 264.33 | |
| 264.33 | |
| Liquid |
| 79271-56-0 | |
| 79°C | |
| MFCD00000407 | |
| 1760 | |
| STMPXDBGVJZCEX-UHFFFAOYSA-N | |
| triethylsilyl trifluoromethanesulfonate | |
| 2733308 | |
| ≥98.0% (T) |
Chemical Identifiers
| 79271-56-0 | |
| 264.33 | |
| STMPXDBGVJZCEX-UHFFFAOYSA-N | |
| 2733308 | |
| CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F |
| C7H15F3O3SSi | |
| MFCD00000407 | |
| triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf | |
| triethylsilyl trifluoromethanesulfonate |
Safety and Handling
TSCA : No