missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilanol 98.0+%, TCI America™

Catalog No. p-7137063
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20175ML 5 mL
T201725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20175ML Supplier TCI America Supplier No. T20175ML
Only null left

Chemical Identifiers

CAS 597-52-4
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00042640
InChI Key WVMSIBFANXCZKT-UHFFFAOYSA-N
Synonym triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
PubChem CID 69005
IUPAC Name triethyl(hydroxy)silane
SMILES CC[Si](CC)(CC)O

Specifications

Color Colorless
Boiling Point 154°C
Quantity 5 mL
UN Number 1987
Formula Weight 132.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylsilanol