missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilanol, 97%

CAS: 597-52-4 | C6H16OSi | 132.278 g/mol

Supplier:  Thermo Scientific Chemicals L0373806

 View more versions of this product

Catalog No. AAL0373806

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

154°C
597-52-4
C6H16OSi
UN1993
57°C (135°F)
WVMSIBFANXCZKT-UHFFFAOYSA-N
triethyl(hydroxy)silane
69005
97%
1.433
Triethylsilanol
5 g
MFCD00042640
1732848
triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
CC[Si](CC)(CC)O
132.278
132.28
Mild
0.86

Chemical Identifiers

597-52-4
132.278
WVMSIBFANXCZKT-UHFFFAOYSA-N
69005
CC[Si](CC)(CC)O
C6H16OSi
MFCD00042640
triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
triethyl(hydroxy)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 209-903-6

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only