missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilane, 98+%

Catalog No. AAA1032030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1032030 250 g
AAA1032009 10 g
AAA1032018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1032030 Supplier Thermo Scientific Chemicals Supplier No. A1032030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-86-7 | C6H15Si | 115.271 g/mol

Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsilane is used for the hydrosilation of olefins to prepare alkyl silanes. It acts as a reducing agent in the reduction of 2-chromanols, since it has an active hydride. It acts as a catalyst for redox initiated cationic polymerization, regioselective reductive coupling of enones and allenes, and Beckmann rearrangement of cyclododecanone oxime. It is associated with trifluoroacetic acid and involved in the selective reduction of alkenes.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, water and bases.

Chemical Identifiers

CAS 617-86-7
Molecular Formula C6H15Si
Molecular Weight (g/mol) 115.271
MDL Number MFCD00009018
InChI Key QXTIBZLKQPJVII-UHFFFAOYSA-N
Synonym triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
PubChem CID 6327258
IUPAC Name triethylsilicon
SMILES CC[Si](CC)CC

Specifications

Boiling Point 107°C to 108°C
Melting Point -157°C
Quantity 250 g
Flash Point −6°C (21°F)
Refractive Index 1.412
Sensitivity Moisture sensitive
Density 0.728
Linear Formula (CH3CH2)3SiH
UN Number UN1993
Beilstein 1098278
Solubility Information Miscible with water.
Formula Weight 116.28
Percent Purity ≥98%
Chemical Name or Material Triethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-535-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only