missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyloxonium tetrafluoroborate, 1M solution in methylene chloride, AcroSeal™
CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol
Supplier: Thermo Scientific Chemicals 429161000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagentGreener Choice Claims
- Environmental benefits include:
- Recyclable
- Renewable Materials
Specifications
Triethyloxonium tetrafluoroborate | |
Colorless to Brown | |
AcroSeal™ Glass bottle | |
(C2H5)3OBF4 | |
100 mL | |
15, 5867 | |
Solubility in water: reacts | |
F[B-](F)(F)F.CC[O+](CC)CC | |
189.99 | |
189.99 |
368-39-8,75-09-2 | |
1.3280g/mL | |
C6H15BF4O | |
MFCD00044423 | |
1.328 | |
triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium | |
IYDQMLLDOVRSJJ-UHFFFAOYSA-N | |
triethyloxidanium;tetrafluoroborate | |
2723982 | |
Liquid |
RUO – Research Use Only