missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium Tetrafluoroborate (15% in Dichloromethane, ca. 1mol/L) [Ethylating Reagent], TCI America™

Catalog No. T1606100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1606100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1606100ML Supplier TCI America Supplier No. T1606100ML

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name tetrafluoroboranuide; triethyloxidanium
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Color Yellow
Quantity 100 mL
UN Number 3265
Formula Weight 189.99
Physical Form Liquid
Chemical Name or Material Triethyloxonium Tetrafluoroborate (15% in Dichloromethane, ca. 1mol/L) [Ethylating Reagent]
TSCA Yes
Recommended Storage Freezer