missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane

Catalog No. p-7081628
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11678AC 25 mL
AAA11678AE 100 mL
AAA11678AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA11678AC Supplier Thermo Scientific Chemicals Supplier No. A11678AC
Only null left

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

Solubility
Soluble in Dichloromethane. Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Density 1.328
Odor Chloroform-like
Linear Formula (CH3CH2)3OBF4
Quantity 25 mL
UN Number UN2922
Beilstein 3598090
Sensitivity Moisture sensitive
Solubility Information Soluble in Dichloromethane. Reacts with water.
Formula Weight 189.99
Concentration or Composition (by Analyte or Components) 1.0M in dichloromethane
Chemical Name or Material Triethyloxonium tetrafluoroborate
Show More Show Less
Hazard Category H314-H336-H350
Hazard Statement GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-705-1
RTECSNumber RR4584700
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only