missing translation for 'onlineSavingsMsg'
Learn More

Triethylmethylammonium Chloride 98.0+%, TCI America™

Catalog No. p-7132405
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M085225G 25 g
M0852500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M085225G Supplier TCI America Supplier No. M085225G
Only null left

Chemical Identifiers

CAS 10052-47-8
Molecular Formula C7H18ClN
Molecular Weight (g/mol) 151.678
MDL Number MFCD00059972
InChI Key NIUZJTWSUGSWJI-UHFFFAOYSA-M
Synonym triethylmethylammonium chloride, n,n-diethyl-n-methylethanaminium chloride, methyltriethylammonium chloride, unii-z0tk7k977q, ethanaminium, n,n-diethyl-n-methyl-, chloride, triethyl methyl azanium chloride, methyl triethyl ammonium chloride, ethanaminium, n,n-diethyl-n-methyl-, chloride 1:1, triethyl-methyl-azanium, acmc-1bu0o
PubChem CID 82326
IUPAC Name triethyl(methyl)azanium;chloride
SMILES CC[N+](C)(CC)CC.[Cl-]

Specifications

Color White
Quantity 25 g
Formula Weight 151.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylmethylammonium Chloride
EINECSNumber (2)-0186
TSCA Yes