missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines

CAS: 112-24-3 | C6H18N4 | 146.238 g/mol

$306.67 - $306.67

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
MDL Number MFCD00008169
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA161280F
View Documents
Thermo Scientific Chemicals
A161280F
2500 mL
Each for $306.67
Only null left
 
Description

Description

Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications

Chemical Identifiers

112-24-3
146.238
VILCJCGEZXAXTO-UHFFFAOYSA-N
5565
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
C6H18N4
MFCD00008169
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
CHEBI:39501
C(CNCCNCCN)N

Specifications

112-24-3
0.982
122°C (252°F)
C6H18N4
H2NCH2CH2(NHCH2CH2)2NH2
2500 mL
605448
14,9663
Insoluble in water.
C(CNCCNCCN)N
146.238
CHEBI:39501
60%
Triethylenetetramine
∼-35°C
∼267°C
Amine-like
1.498
MFCD00008169
UN2259
Hygroscopic
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
VILCJCGEZXAXTO-UHFFFAOYSA-N
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
5565
146.24
Technical
balance branched and cyclic triethylenetetramines
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H312-H317
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H332-H311-H314-H317-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRIETHYLENETETRAMINE

EINECSNumber : 203-950-6

RTECSNumber : YE6650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only